Learn More
Medchemexpress LLC Givinostat hydrochloride | 199657-29-9 | 98.06% | 457.95 | 25 MG
SDP
Catalog No. 5000362770
Encompass_Preferred
Givinostat hydrochloride (ITF-2357) is a potent Histone Deacetylase (HDAC) inhibitor, with IC50 values of 198 nM for HDAC1 and 157 nM for HDAC3. This compound can also penetrate the blood-brain barrier (BBB) and is typically a white to off-white solid.
  • Potent HDAC inhibitor
  • Penetrates the blood-brain barrier
  • White to off-white solid appearance
  • Store solid at 4°C, sealed, away from moisture
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-62770 Supplier Medchemexpress LLC Supplier No. 25MGHY14842A
Givinostat hydrochloride (ITF-2357) is a potent Histone Deacetylase (HDAC) inhibitor, with IC50 values of 198 nM for HDAC1 and 157 nM for HDAC3. This compound can also penetrate the blood-brain barrier (BBB) and is typically a white to off-white solid.
  • Potent HDAC inhibitor
  • Penetrates the blood-brain barrier
  • White to off-white solid appearance
  • Store solid at 4°C, sealed, away from moisture
  • Store in solvent at -80°C for 6 months or -20°C for 1 month