Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Givinostat (hydrochloride) | 199657-29-9 | 98.1% | 457.95 | 1 ML
SDP

Catalog No. 5000362076
Encompass_Preferred

Givinostat (ITF-2357) hydrochloride is an HDAC inhibitor that can penetrate the blood-brain barrier (BBB). It demonstrates inhibitory activity against various HDACs, including HDAC1 (IC50: 198 nM) and HDAC3 (IC50: 157 nM).

  • Targets multiple HDACs, including hHDAC1, hHDAC3, hHDAC11, hHDAC6, hHDAC2, hHDAC10, hHDAC7, hHDAC5, hHDAC9, hHDAC8, and hHDAC4.
  • Suppresses LPS-induced IL-1β production by over 70%.
  • Reduces IL-6 production in PBMCs stimulated with TLR agonists.
  • Inhibits JS-1 cell proliferation.
  • Reduces serum TNFα in vivo.

Catalog No. 50-003-62076 Supplier Medchemexpress LLC Supplier No. HY14842A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Givinostat (ITF-2357) hydrochloride is an HDAC inhibitor that can penetrate the blood-brain barrier (BBB). It demonstrates inhibitory activity against various HDACs, including HDAC1 (IC50: 198 nM) and HDAC3 (IC50: 157 nM).

  • Targets multiple HDACs, including hHDAC1, hHDAC3, hHDAC11, hHDAC6, hHDAC2, hHDAC10, hHDAC7, hHDAC5, hHDAC9, hHDAC8, and hHDAC4.
  • Suppresses LPS-induced IL-1β production by over 70%.
  • Reduces IL-6 production in PBMCs stimulated with TLR agonists.
  • Inhibits JS-1 cell proliferation.
  • Reduces serum TNFα in vivo.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.