Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Givinostat hydrochloride | 199657-29-9 | MFCD28502062 | 98.1% | 457.95 | 100 MG
SDP

Catalog No. 5000362077
Encompass_Preferred

Givinostat hydrochloride is a Histone Deacetylase (HDAC) inhibitor that can penetrate the blood-brain barrier (BBB). It is known to inhibit HDAC1 and HDAC3, and has shown efficacy in suppressing inflammatory responses and inhibiting cell proliferation in various studies.

  • Inhibits HDAC1 (IC50: 198 nM) and HDAC3 (IC50: 157 nM)
  • Penetrates the blood-brain barrier (BBB)
  • Suppresses LPS-induced IL-1β production
  • Decreases IL-6 secretion in PBMCs
  • Inhibits JS-1 cell proliferation

Catalog No. 50-003-62077 Supplier Medchemexpress LLC Supplier No. HY14842A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Givinostat hydrochloride is a Histone Deacetylase (HDAC) inhibitor that can penetrate the blood-brain barrier (BBB). It is known to inhibit HDAC1 and HDAC3, and has shown efficacy in suppressing inflammatory responses and inhibiting cell proliferation in various studies.

  • Inhibits HDAC1 (IC50: 198 nM) and HDAC3 (IC50: 157 nM)
  • Penetrates the blood-brain barrier (BBB)
  • Suppresses LPS-induced IL-1β production
  • Decreases IL-6 secretion in PBMCs
  • Inhibits JS-1 cell proliferation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.