Learn More
TARGETMOL CHEMICALS INC GIVINOSTAT HYDROCHLORIDE 5MG
SDP
Catalog No. 502579457
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Givinostat hydrochloride (ITF2357 hydrochloride) is an inhibitor of HDAC with IC50 of 198 and 157 nM for HDAC1 and HDAC3 respectively. Givinostat hydrochloride exhibits anti-inflammatory anti-angiogenic and antineoplastic activities. purity: 98%
Catalog No. 50-257-9457 Supplier TARGETMOL CHEMICALS INC Supplier No. T6279L5MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Givinostat hydrochloride (ITF2357 hydrochloride) is an inhibitor of HDAC with IC50 of 198 and 157 nM for HDAC1 and HDAC3 respectively. Givinostat hydrochloride exhibits anti-inflammatory anti-angiogenic and antineoplastic activities. purity: 98%