Learn More
Medchemexpress LLC Givinostat hydrochloride monohydrate | 732302-99-7 | 99.6% | 475.97 | 100 MG
SDP
Catalog No. 5000361608
Encompass_Preferred
Givinostat hydrochloride monohydrate is a potent histone deacetylase (HDAC) inhibitor, demonstrating inhibitory activity against HDAC1 (IC50 of 198 nM) and HDAC3 (IC50 of 157 nM). It can penetrate the blood-brain barrier (BBB), making it suitable for neurological research.
  • Potent HDAC inhibitor targeting HDAC1 and HDAC3.
  • Can penetrate the blood-brain barrier (BBB).
  • Suitable for in vitro and in vivo studies related to HDAC inhibition.
  • Available as a stable hydrochloride monohydrate form.
Catalog No. 50-003-61608 Supplier Medchemexpress LLC Supplier No. HY14842B100MG
Givinostat hydrochloride monohydrate is a potent histone deacetylase (HDAC) inhibitor, demonstrating inhibitory activity against HDAC1 (IC50 of 198 nM) and HDAC3 (IC50 of 157 nM). It can penetrate the blood-brain barrier (BBB), making it suitable for neurological research.
  • Potent HDAC inhibitor targeting HDAC1 and HDAC3.
  • Can penetrate the blood-brain barrier (BBB).
  • Suitable for in vitro and in vivo studies related to HDAC inhibition.
  • Available as a stable hydrochloride monohydrate form.