Learn More
Medchemexpress LLC Givinostat hydrochloride monohydrate | 732302-99-7 | 99.6% | 475.97 | 25 MG
SDP
Catalog No. 5000363756
Encompass_Preferred
Givinostat hydrochloride monohydrate is a potent HDAC inhibitor, demonstrating IC50 values of 198 nM for HDAC1 and 157 nM for HDAC3. This compound is notable for its ability to penetrate the blood-brain barrier, making it suitable for a range of research applications.
  • Potent HDAC inhibitor for HDAC1 and HDAC3
  • Penetrates the blood-brain barrier
  • Suppresses LPS-induced IL-1β and IL-6 production
  • Inhibits JS-1 cell proliferation
  • Reduces serum TNFα and IL-1β levels in vivo
Catalog No. 50-003-63756 Supplier Medchemexpress LLC Supplier No. 25MGHY14842B
Givinostat hydrochloride monohydrate is a potent HDAC inhibitor, demonstrating IC50 values of 198 nM for HDAC1 and 157 nM for HDAC3. This compound is notable for its ability to penetrate the blood-brain barrier, making it suitable for a range of research applications.
  • Potent HDAC inhibitor for HDAC1 and HDAC3
  • Penetrates the blood-brain barrier
  • Suppresses LPS-induced IL-1β and IL-6 production
  • Inhibits JS-1 cell proliferation
  • Reduces serum TNFα and IL-1β levels in vivo