Learn More
Medchemexpress LLC GJ071 oxalate | 1216676-34-4 | 99.7% | 455.53 | 1 MG
SDP
Catalog No. 5000348578
Encompass_Preferred
GJ071 oxalate is a chemical that induces ATM kinase activity in ataxia telangiectasia (A-T) cells carrying homozygous TGA or TAG stop codons. It is intended for research use only and is not for patient use.
  • Induces ATM kinase activity
  • White to off-white solid appearance
  • Shipping at room temperature (continental US)
  • Storage at -20°C (sealed, away from moisture)
  • For research use only
Catalog No. 50-003-48578 Supplier Medchemexpress LLC Supplier No. HY1368351MG
GJ071 oxalate is a chemical that induces ATM kinase activity in ataxia telangiectasia (A-T) cells carrying homozygous TGA or TAG stop codons. It is intended for research use only and is not for patient use.
  • Induces ATM kinase activity
  • White to off-white solid appearance
  • Shipping at room temperature (continental US)
  • Storage at -20°C (sealed, away from moisture)
  • For research use only