Learn More
Medchemexpress LLC GJ071 oxalate | 1216676-34-4 | 99.7% | 455.53 g/mol | C20H29N3O7S | 5 MG
SDP
Catalog No. 5000222439
Encompass_Preferred
GJ071 oxalate is a research-grade small molecule reported to induce ATM kinase activity in ataxia telangiectasia (A-T) cells carrying homozygous TGA or TAG stop codons. Supplied as the oxalate salt for laboratory research use.
  • Induces ATM kinase activity in A-T cells with TGA or TAG stop codons.
  • High purity: 99.74%.
  • Molecular formula: C20H29N3O7S; molecular weight: 455.53 g/mol.
  • Available in small-scale laboratory quantities (1 mg-100 mg).
  • Suitable for cellular research applications; soluble in DMSO.
Catalog No. 50-002-22439 Supplier Medchemexpress LLC Supplier No. HY1368355MG
GJ071 oxalate is a research-grade small molecule reported to induce ATM kinase activity in ataxia telangiectasia (A-T) cells carrying homozygous TGA or TAG stop codons. Supplied as the oxalate salt for laboratory research use.
  • Induces ATM kinase activity in A-T cells with TGA or TAG stop codons.
  • High purity: 99.74%.
  • Molecular formula: C20H29N3O7S; molecular weight: 455.53 g/mol.
  • Available in small-scale laboratory quantities (1 mg-100 mg).
  • Suitable for cellular research applications; soluble in DMSO.