Learn More
Medchemexpress LLC GJ071 oxalate | 1216676-34-4 | 99.7% | C20H29N3O7S | 10MG
SDP
Catalog No. 5000260206
Encompass_Preferred
GJ071 oxalate is a research-grade small molecule that induces ATM kinase activity in ataxia telangiectasia (A-T) cells carrying homozygous TGA or TAG stop codons. Supplied as a solid with high reported purity, it is intended for in vitro biochemical and cellular studies; follow supplier handling and storage recommendations.
Catalog No. 50-002-60206 Supplier Medchemexpress LLC Supplier No. HY13683510MG
GJ071 oxalate is a research-grade small molecule that induces ATM kinase activity in ataxia telangiectasia (A-T) cells carrying homozygous TGA or TAG stop codons. Supplied as a solid with high reported purity, it is intended for in vitro biochemical and cellular studies; follow supplier handling and storage recommendations.