Learn More
Medchemexpress LLC GJ103 sodium salt | 1459687-96-7 | 99.5% | 364.35 g/mol | C16H13N4NaO3S | 25 MG
SDP
Catalog No. 5000211200
Encompass_Preferred
GJ103 sodium is a small-molecule read-through compound (sodium salt) that promotes ribosomal read-through of premature stop codons and is used in research on genetic disorders caused by nonsense mutations. It is supplied as a high-purity solid for use in biochemical, cellular, and exploratory in vivo studies; handle and store per supplier recommendations.
Catalog No. 50-002-11200 Supplier Medchemexpress LLC Supplier No. HY101203A25MG
GJ103 sodium is a small-molecule read-through compound (sodium salt) that promotes ribosomal read-through of premature stop codons and is used in research on genetic disorders caused by nonsense mutations. It is supplied as a high-purity solid for use in biochemical, cellular, and exploratory in vivo studies; handle and store per supplier recommendations.