Learn More
Aviva Systems Biology Corp Gja10 Antibody - N-terminal region (ARP42834_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42834_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Gja10. It was validated on Western Blot. Gja10,gap junction protein, alpha 10,Cx59; Cx-57; Gja10,gap junction alpha-10 protein,gap junction alpha-10 protein,cx57,connexin-57,OTTMUSP00000005122,gap junction membrane channel protein alpha 10The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Cx-57, Cx59ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-033