Learn More
Aviva Systems Biology Corp GJA3 Antibody - N-terminal region (ARP36626_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36626_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GJA3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GJA3. Alternate names: CX46, CZP3ApplicationWB. Species reactivity: Horse, Human, Mouse, Pig, Rat, Yeast, Zebrafish
Catalog No. 50-014-56218