Learn More
Aviva Systems Biology Corp GJA5 Antibody - N-terminal region (ARP36610_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36610_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GJA5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GJA5. Alternate names: CX40, ATFB11ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-53055