Learn More
Aviva Systems Biology Corp GJA9 Antibody - middle region (ARP36628_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36628_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GJA9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GJA9. Alternate names: CX58, CX59, GJA10, MGC50985ApplicationWB. Species reactivity: Dog, Human
Catalog No. 50-014-56219