Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 2705MSM3P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: GJB1/1753. Clonality: Monoclonal. Host: Mouse. Isotype: IgG. Positive Control: MCF-7 Cells. Liver, Kidney, Stomach or Tonsil. Cellular Staining: Cytoplasmic and Cell Surface. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human GJB1 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 27-32kDa.Chromosome: Xq13.1. Gene Symbol: GJB1. Gene ID: 2705. Ensemble: ENSG00000169562. Uniprot: XP_016884897.
Catalog No. 50-188-4735