Learn More
Aviva Systems Biology Corp GJB1 Antibody - middle region (ARP36599_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36599_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GJB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GJB1. Alternate names: CMTX, CMTX1, CX32ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56208