Learn More
Aviva Systems Biology Corp Gjb3 Antibody - N-terminal region (ARP42822_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42822_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Gjb3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Cnx31, Cx31, D4Wsu144e, Gjb-3ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-030