Learn More
Aviva Systems Biology Corp GJB6 Antibody - middle region (ARP36617_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36617_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GJB6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GJB6. Alternate names: CX30, DFNA3, ED2, EDH, HED, DFNA3B, DFNB1BApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56214