Learn More
Aviva Systems Biology Corp GJC3 Antibody - middle region (ARP36639_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36639_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GJC3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human GJC3. Alternate names: CX29, CX30.2, CX31.3, GJE1ApplicationWB. Species reactivity: Human
Catalog No. 50-284-507