Learn More
Aviva Systems Biology Corp GJD2 Antibody - middle region (ARP36623_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36623_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GJD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GJD2. Alternate names: CX36, GJA9, MGC138315, MGC138319ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56217