Learn More
Aviva Systems Biology Corp GJE1 Antibody - C-terminal region (ARP36638_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36638_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GJE1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human GJE1. Alternate names: CX29, GJE1, CX30.2, CX31.3ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56225