Learn More
Aviva Systems Biology Corp GK Antibody - middle region (ARP55955_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55955_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GK. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human GK. Alternate names: GK1, GKDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-099