Learn More
ATCC GK1.5; Hybridoma; Rat/Mouse (Rattus norvegicus/Mus musculus)
Catalog No. 502381776
Encompass
GK1.5 is a B lymphocyte cell that was isolated from the spleen of a mouse with leukemia. The antibody profoundly blocks antigen specific murine class II MHC antigen reactive helper T lymphocyte lines.
Catalog No. 50-238-1776 Supplier ATCC Supplier No. TIB207
GK1.5 is a B lymphocyte cell that was isolated from the spleen of a mouse with leukemia. The antibody profoundly blocks antigen specific murine class II MHC antigen reactive helper T lymphocyte lines.