Learn More
Aviva Systems Biology Corp GK2 Antibody - N-terminal region (ARP53814_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53814_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GK2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human GK2. Alternate names: GKP2, GKTAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-892