Learn More
Aviva Systems Biology Corp GK2 Antibody - C-terminal region (ARP53815_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53815_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GK2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human GK2. Alternate names: GKP2, GKTAApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Mouse, Rabbit
Catalog No. 50-014-61136