Learn More
Selleck Chemical LLC GK921
SDP
Catalog No. 5000002989
Encompass_Preferred
GK921 is a transglutaminase 2 (TGase 2) inhibitor with an IC50 of 8 93 M under a modified assay condition
Catalog No. 50-000-02989 Supplier Selleck Chemical LLC Supplier No. S64172MG
GK921 is a transglutaminase 2 (TGase 2) inhibitor with an IC50 of 8 93 M under a modified assay condition