Learn More
Medchemexpress LLC HY-100521 5mg Medchemexpress, GKI-1 CAS: Purity:>98%
SDP
Catalog No. 501967737
Encompass_Preferred
Medchemexpress, HY-100521 5mg GKI-1 CAS: GKI-1 is a Greatwall (GWL) kinase inhibitor with IC50s of 4.9 and 2.5 µM against hGWLFL and hGWL-KinDom, respectively. GKI-1 robustly inhibits ROCK1 with an IC50 of 11 µM, but only weakly affected PKA. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7737 Supplier Medchemexpress LLC Supplier No. HY1005215MG
Medchemexpress, HY-100521 5mg GKI-1 CAS: GKI-1 is a Greatwall (GWL) kinase inhibitor with IC50s of 4.9 and 2.5 µM against hGWLFL and hGWL-KinDom, respectively. GKI-1 robustly inhibits ROCK1 with an IC50 of 11 µM, but only weakly affected PKA. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.