Learn More
TARGETMOL CHEMICALS INC GKI-1 HCL 5MG
SDP
Catalog No. 502581533
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GKI-1 HCl is a Greatwall (GWL) kinase inhibitor that inhibits hGWLFL and hGWL-KinDom.The inhibitory effect of GKI-1 HCl on ROCK1 is more significant.
Catalog No. 50-258-1533 Supplier TARGETMOL CHEMICALS INC Supplier No. T11402L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GKI-1 HCl is a Greatwall (GWL) kinase inhibitor that inhibits hGWLFL and hGWL-KinDom.The inhibitory effect of GKI-1 HCl on ROCK1 is more significant.