Learn More
Medchemexpress LLC HY-101499 5mg , GKT136901 CAS:955272-06-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1014995MG
Encompass_Preferred
HY-101499 5mg GKT136901 CAS: 955272-06-7 GKT136901 is a potent, selective and orally active inhibitor of NADPH oxidase (NOX1/4), with Kis of 160 and 165 nM, respectively. GKT136901 is also a selective and direct scavenger of peroxynitrite. GKT136901 can be used for the research of diabetic nephropathy, stroke, and neurodegeneration. GKT136901 also has anti-inflammatory activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1251