Learn More
Medchemexpress LLC GKT136901 hydrochloride | 1254507-01-1 | 99.9% | 403.26 g/mol | C19H16Cl2N4O2 | 1 ML
SDP
Catalog No. 5000248790
Encompass_Preferred
GKT136901 hydrochloride is a research-grade small-molecule inhibitor selective for NADPH oxidase isoforms NOX1 and NOX4. It is used in preclinical studies to investigate oxidative stress, inflammation, and related disease models such as diabetic nephropathy, stroke, and neurodegeneration. The hydrochloride salt is supplied as a solid or as a 10 mM solution in DMSO for biochemical and cell-based assays.
  • Selective NOX1/4 inhibition for targeted research applications.
  • Potent activity (Ki ≈ 160 nM) for effective pathway modulation.
  • Available as a 10 mM solution in DMSO for ready-to-use assays.
  • High chemical purity to support reproducible results.
  • Supplied as a stable solid with recommended cold storage.
  • Suitable for biochemical and cell-based experimental workflows.
Catalog No. 50-002-48790 Supplier Medchemexpress LLC Supplier No. HY101499A10MM 1ML
GKT136901 hydrochloride is a research-grade small-molecule inhibitor selective for NADPH oxidase isoforms NOX1 and NOX4. It is used in preclinical studies to investigate oxidative stress, inflammation, and related disease models such as diabetic nephropathy, stroke, and neurodegeneration. The hydrochloride salt is supplied as a solid or as a 10 mM solution in DMSO for biochemical and cell-based assays.
  • Selective NOX1/4 inhibition for targeted research applications.
  • Potent activity (Ki ≈ 160 nM) for effective pathway modulation.
  • Available as a 10 mM solution in DMSO for ready-to-use assays.
  • High chemical purity to support reproducible results.
  • Supplied as a stable solid with recommended cold storage.
  • Suitable for biochemical and cell-based experimental workflows.