Learn More
Medchemexpress LLC Gkt136901 (hydrochloride) | 1254507-01-1 | 99.9% | 403.26 g/mol | C19H16Cl2N4O2 | 50 MG
SDP
Catalog No. 5000247942
Encompass_Preferred
GKT136901 hydrochloride is the hydrochloride salt of a selective NADPH oxidase (NOX1/4) inhibitor used in research on oxidative stress-related disease models such as diabetic nephropathy, stroke, and neurodegeneration. It is supplied as a solid with defined storage conditions and formulation recommendations for reliable experimental use.
  • Selective NOX1/4 inhibition for oxidative stress research.
  • High purity suitable for analytical and biological studies.
  • Available in multiple small pack sizes for lab-scale experiments.
  • Stable as a solid when stored sealed at 4°C.
  • Soluble in DMSO for solution preparations; 10 mM recommended.
Catalog No. 50-002-47942 Supplier Medchemexpress LLC Supplier No. HY101499A50MG
GKT136901 hydrochloride is the hydrochloride salt of a selective NADPH oxidase (NOX1/4) inhibitor used in research on oxidative stress-related disease models such as diabetic nephropathy, stroke, and neurodegeneration. It is supplied as a solid with defined storage conditions and formulation recommendations for reliable experimental use.
  • Selective NOX1/4 inhibition for oxidative stress research.
  • High purity suitable for analytical and biological studies.
  • Available in multiple small pack sizes for lab-scale experiments.
  • Stable as a solid when stored sealed at 4°C.
  • Soluble in DMSO for solution preparations; 10 mM recommended.