Learn More
Selleck Chemical LLC Setanaxib (GKT137831) 5mg 1218942-37-0
SDP
Supplier:  Selleck Chemical LLC S71715MG
Encompass_Preferred
Setanaxib (GKT137831) is a potent, dual NADPH oxidase NOX1/NOX4 inhibitor with Ki of 110 nM and 140 nM, respectively. Treatment with GKT137831 suppresses reactive oxygen species (ROS) production. GKT137831 partly inhibits ferroptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1012703