Learn More
Chemglass Life Sciences GL-14 Solid Cap
SDP
Catalog No. 501531945
Encompass_Preferred
GL-14 Solid Cap. Cap is made from PBTE (thermoplastic polyester) and will withstand temperatures from -45 to 200 C. PTFE seal is available separately.
Catalog No. 50-153-1945 Supplier Chemglass Life Sciences Supplier No. CGB19601
GL-14 Solid Cap. Cap is made from PBTE (thermoplastic polyester) and will withstand temperatures from -45 to 200 C. PTFE seal is available separately.