Learn More
Chemglass Life Sciences GL-18 Cap with Aperture, High Temperature
SDP
Catalog No. CG195H02
Encompass_Preferred
GL-18 Cap with Aperture, High Temperature. Open top cap for use with GL screw threads. The cap combined with the CG-197 sealing ring will accommodate tubing of the listed diameter. Cap is made from PPS (polyphenylsiloxan) and will withstand temperatures up to 250°C.
Catalog No. CG195H02 Supplier Chemglass Life Sciences Supplier No. CG195H02
GL-18 Cap with Aperture, High Temperature. Open top cap for use with GL screw threads. The cap combined with the CG-197 sealing ring will accommodate tubing of the listed diameter. Cap is made from PPS (polyphenylsiloxan) and will withstand temperatures up to 250°C.