Learn More
Chemglass Life Sciences GL-18 Cap with 11mm Aperture
SDP
Supplier:  Chemglass Life Sciences CGB19502
Encompass_Preferred
GL-18 Cap with 11mm Aperture. Open top cap for use with GL screw threads. The cap combined with the CG-197 sealing ring will accommodate tubing of the listed diameter. Cap is made from PBTE (thermoplastic polyester) and will withstand temperatures from -45 to 200 C.
Catalog No. 50-153-1941