Learn More
Chemglass Life Sciences GL-18 Solid Cap
SDP
Catalog No. 501531946
Encompass_Preferred
GL-18 Solid Cap. Cap is made from PBTE (thermoplastic polyester) and will withstand temperatures from -45 to 200 C. PTFE seal is available separately.
Catalog No. 50-153-1946 Supplier Chemglass Life Sciences Supplier No. CGB19602
GL-18 Solid Cap. Cap is made from PBTE (thermoplastic polyester) and will withstand temperatures from -45 to 200 C. PTFE seal is available separately.