Learn More
Chemglass Life Sciences GL-25 Cap with 15mm Aperture
SDP
Supplier:  Chemglass Life Sciences CG19503
Encompass_Preferred
GL-25 Cap with 15mm Aperture. Open top cap for use with GL screw threads. The cap combined with the CG-197 sealing ring will accommodate tubing of the listed diameter. Cap is made from PBT (thermoplastic polyester) and will withstand temperatures from -45 to 200º C.
Catalog No. CG19503