Learn More
Chemglass Life Sciences GL-25 Screw Thread for Thermocouple Bushing
SDP
Catalog No. 501531634
Encompass_Preferred
GL-25 Screw Thread for Thermocouple Bushing. Connector allows for easy connection of a thermocouple gauge tube to glass vacuum system without the use of a glass to metal graded seal or Cajon fitting. Gauge connection consists of a GL 25 thread, a Stainless Steel bushing and an open top GL 25 screw cap. The thermocouple gauge tube is threaded into the #303 Stainless Steel bushing and inserted into the glass GL 25 thread.
Catalog No. 50-153-1634 Supplier Chemglass Life Sciences Supplier No. CG35411
GL-25 Screw Thread for Thermocouple Bushing. Connector allows for easy connection of a thermocouple gauge tube to glass vacuum system without the use of a glass to metal graded seal or Cajon fitting. Gauge connection consists of a GL 25 thread, a Stainless Steel bushing and an open top GL 25 screw cap. The thermocouple gauge tube is threaded into the #303 Stainless Steel bushing and inserted into the glass GL 25 thread.