Learn More
Chemglass Life Sciences GL-32 Cap with 20.0mm Aperture
SDP
Catalog No. 501531943
Encompass_Preferred
GL-32 Cap with 20.0mm Aperture. Open top cap for use with GL screw threads. The cap combined with the CG-197 sealing ring will accommodate tubing of the listed diameter. Cap is made from PBTE (thermoplastic polyester) and will withstand temperatures from -45 to 200 C.
Catalog No. 50-153-1943 Supplier Chemglass Life Sciences Supplier No. CGB19504
GL-32 Cap with 20.0mm Aperture. Open top cap for use with GL screw threads. The cap combined with the CG-197 sealing ring will accommodate tubing of the listed diameter. Cap is made from PBTE (thermoplastic polyester) and will withstand temperatures from -45 to 200 C.