Learn More
Chemglass Life Sciences GL-32 Screw Thread, 28mm X 2.0W, 100mm OAL
SDP
Catalog No. 501531938
Encompass_Preferred
GL-32 Screw Thread, 28mm X 2.0W, 100mm OAL. Screw thread tube for replacement on apparatus using the GL thread, which is manufactured to I.S.O. specifications. (GL is the I.S.O. designation for GLASS.) To be used with a CG-196 solid cap with liner or a CG-195 cap with aperture and CG-197 silicone sealing ring.
Catalog No. 50-153-1938 Supplier Chemglass Life Sciences Supplier No. CGB19404
GL-32 Screw Thread, 28mm X 2.0W, 100mm OAL. Screw thread tube for replacement on apparatus using the GL thread, which is manufactured to I.S.O. specifications. (GL is the I.S.O. designation for GLASS.) To be used with a CG-196 solid cap with liner or a CG-195 cap with aperture and CG-197 silicone sealing ring.