Learn More
Analytical Sales & Services GL-38 Bottle Cap - (2) 3/16" OD Tubing Ports
Catalog No. 5000804405
Encompass
Mobile Phase Bottle Cap for GL-38 4L Bottles - (2) 3/16" OD Tubing Ports\nColor: Black\n\nQty/Pkg: EACH
Catalog No. 50-008-04405 Supplier Analytical Sales & Services Supplier No. 38316
Add to Cart
Add to Cart
Mobile Phase Bottle Cap for GL-38 4L Bottles - (2) 3/16" OD Tubing Ports\nColor: Black\n\nQty/Pkg: EACH