Learn More
Chemglass Life Sciences Transfer Assembly, 125mL, 2-Port, GL-45 Cap, Advantaflex Tubing, Sterile
SDP
Catalog No. 501979499
Encompass
Chemglass Life Sciences GL-45 cap transfer assemblies are available in sizes from 125mL-1L.These transfer assemblies are extremely helpful for sterile transfer of liquids from containers or bottles which have a GL-45 threaded neck.Our assemblies are a great choice to achieve sterile fluid transfer when using Corning and Thermo Fisher filter systems and storage bottles.Each 2-port GL-45 closure assembly includes a 0. 22um filter, 24-inch length of Advantaflex tubing with a plugged end, 1/4” pinch clamp and a V- notched dip tube.The assembly is made from pharmaceutical grade thermoplastic elastomer resin.Manufactured, assembled and packaged in a Class 7 cleanroom, USP Class VI approved components.Each assembly is individually double poly-bagged and gamma-irradiated.Please note: Compatable with Fisher/Nunc brand and Corning brand filter media bottles.
Catalog No. 50-197-9499 Supplier Chemglass Life Sciences Supplier No. CLS11630125
Chemglass Life Sciences GL-45 cap transfer assemblies are available in sizes from 125mL-1L.These transfer assemblies are extremely helpful for sterile transfer of liquids from containers or bottles which have a GL-45 threaded neck.Our assemblies are a great choice to achieve sterile fluid transfer when using Corning and Thermo Fisher filter systems and storage bottles.Each 2-port GL-45 closure assembly includes a 0. 22um filter, 24-inch length of Advantaflex tubing with a plugged end, 1/4” pinch clamp and a V- notched dip tube.The assembly is made from pharmaceutical grade thermoplastic elastomer resin.Manufactured, assembled and packaged in a Class 7 cleanroom, USP Class VI approved components.Each assembly is individually double poly-bagged and gamma-irradiated.Please note: Compatable with Fisher/Nunc brand and Corning brand filter media bottles.