Learn More
Ace Glass, Inc. GL45 PBT CAP W/CENTER HOLE
SDP
Catalog No. NC0174504
Encompass
GL45 Duran, polybutylene terephthalate (PBT) open-top caps. PBT temperature range is -45 - 180°C. Can be used with Duran hose connectors, Duran sealing ring for GL thread hose connectors, and silicone or silicone/PTFE septa for Duran caps.
Catalog No. NC0174504 Supplier Ace Glass, Inc. Supplier No. 762125
GL45 Duran, polybutylene terephthalate (PBT) open-top caps. PBT temperature range is -45 - 180°C. Can be used with Duran hose connectors, Duran sealing ring for GL thread hose connectors, and silicone or silicone/PTFE septa for Duran caps.