Learn More
Chemglass Life Sciences GL-45 Cap with 34.0mm Aperture
SDP
Catalog No. 501531944
Encompass_Preferred
GL-45 Cap with 34.0mm Aperture. Open top cap for use with GL screw threads. The cap combined with the CG-197 sealing ring will accommodate tubing of the listed diameter. Cap is made from PBTE (thermoplastic polyester) and will withstand temperatures from -45 to 200 C.
Catalog No. 50-153-1944 Supplier Chemglass Life Sciences Supplier No. CGB19505
GL-45 Cap with 34.0mm Aperture. Open top cap for use with GL screw threads. The cap combined with the CG-197 sealing ring will accommodate tubing of the listed diameter. Cap is made from PBTE (thermoplastic polyester) and will withstand temperatures from -45 to 200 C.