Learn More
Research Products International Corp GL 45 Polypropylene Color Coded Caps, Fits KimCote Bottles and Erlenmeyer Flasks, Blue, 10 per Case
SDP
Supplier:  Research Products International Corp 218205
Encompass
Solid top polypropylene GL 45 screw thread caps have internal molded seal rings. Caps can be used with KimCote Media Bottles and Erlenmeyer Flasks. These caps are autoclavable.
Catalog No. 50-197-8275