Learn More
General Laboratory Products GL TBES W/THDEND RNDBTM 250/CS
Catalog No. 50855959
Encompass_Preferred
Test Tube; Disposable; Borosilicate Glass; Round Bottom; Caps: None; 18-415 Threaded End; O.D. x L: 20 x 150mm
Catalog No. 50-855-959 Supplier General Laboratory Products Supplier No. GLP20150RB
Test Tube; Disposable; Borosilicate Glass; Round Bottom; Caps: None; 18-415 Threaded End; O.D. x L: 20 x 150mm