Learn More
Across International Ai GL18 Chilling Fluid or Vacuum Connectors - Pack of 5
SDP
Catalog No. NC2131578
Encompass_Preferred
GL18 chilling fluid or vacuum connector with sealing gasket, pack of 5. Fits our SolventVap rotary evaporators and glass jacketed reactors.
This item is not returnable. View return policy
Catalog No. NC2131578 Supplier Across International Supplier No. GL18X5
GL18 chilling fluid or vacuum connector with sealing gasket, pack of 5. Fits our SolventVap rotary evaporators and glass jacketed reactors.