Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transforming Technologies GL2500P - ESD Level 3 Cut Resistant Gloves - Palm Coated
SDP

Catalog No. 17226952
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-226-952 X-Small
17-226-953 Small
17-226-955 Medium
17-226-956 Large
17-226-957 X-Large
17-226-958 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-226-952 Supplier Transforming Technologies Supplier No. GL2501P
Add to Cart
Add to Cart

The ESD Cut Resistant Gloves combine powerful cut resistance, comfort and uniform ESD protection into one superior performing glove.

  • Fabric: 18 Gauge Yarn, UHMWPE Construction with Conductive Carbon
  • Uncoated Finish for Max Breathability
  • Cuff: Finished Knitwrist Cuff
  • Sold in packs of 12, 1 size per pack
  • Typical ESD Properties: <107 Ohms
  • Cut Resistant (ANSI Cut Level 3)
  • Semiconductor, SMT Assembly, Disk Drive
  • Applications that require handling of ESD sensitive items while there is a minor cut risk

Specifications

Coating Material Polyurethane
Color Gray
Hand Ambidextrous
Material Composite Yarn
Product Type Cut-resistant Gloves
Size X-Small
Gender Unisex
Packaging Type Box
Quantity 12 pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.