Learn More
Transforming Technologies GL2500T - ESD Level 3 Cut Resistant Gloves - Fingertip Coated
SDP
Catalog No. 17226964
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-226-964 2X-Large
17-226-959 X-Small
17-226-960 Small
17-226-961 Medium
17-226-962 Large
17-226-963 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-226-964 Supplier Transforming Technologies Supplier No. GL2506T
Add to Cart
Add to Cart
The ESD Cut Resistant Gloves combine powerful cut resistance, comfort and uniform ESD protection into one superior performing glove.
  • Fabric: 18 Gauge Yarn, UHMWPE Construction with Conductive Carbon.
  • Uncoated Finish for Max Breathability
  • Cuff: Finished Knitwrist Cuff.
  • Sold in packs of 12, 1 size per pack.
  • Typical ESD Properties: <107 Ohms
  • Cut Resistant (ANSI Cut Level 3)
  • Semiconductor, SMT Assembly, Disk Drive. Applications that require handling of ESD sensitive items while there is a minor cut risk.
Specifications
Coating Material Polyurethane
Color Gray
Hand Ambidextrous
Material Composite Yarn
Product Type Cut-resistant Gloves
Size 2X-Large
Gender Unisex
Packaging Type Box
Puncture Rating ANSI/ISEA 105-16 Level 3
Quantity 12 pack