Learn More
Chemglass Life Sciences Flask, Shake, 1000mL, 6 Baffles, GL-45 Thread, Vented Cap, Approx OD x Height (mm): 130 x 215
SDP
Catalog No. 501214764
Encompass_Preferred
Flask, Shake, 1000mL, 6 Baffles, GL-45 Thread, Vented Cap, Approx OD x Height (mm): 130 x 215. Flask has 6 baffles, 3 baffles on the lower outside edge of the flask and 3 baffles on the bottom surface of the flask to maximize oxygen transfer to the culture medium when used on an orbital shaker. Gray polypropylene vented cap has a 0.22µm (PTFE) hydrophobic membrane to allow gas exchange while preventing contamination. They have a liner-less plug seal design for efficient sealing. They are autoclavable at 121°C. Since these caps are vented, the flasks can be autoclaved with the cap attached.
Catalog No. 50-121-4764 Supplier Chemglass Life Sciences Supplier No. CLS205208
Flask, Shake, 1000mL, 6 Baffles, GL-45 Thread, Vented Cap, Approx OD x Height (mm): 130 x 215. Flask has 6 baffles, 3 baffles on the lower outside edge of the flask and 3 baffles on the bottom surface of the flask to maximize oxygen transfer to the culture medium when used on an orbital shaker. Gray polypropylene vented cap has a 0.22µm (PTFE) hydrophobic membrane to allow gas exchange while preventing contamination. They have a liner-less plug seal design for efficient sealing. They are autoclavable at 121°C. Since these caps are vented, the flasks can be autoclaved with the cap attached.